1-(Methoxymethyl)pyrimidine-2,4(1H,3H)-dione 97%

Supplier: Ambeed
Warning

A1483868-100MG A1483868-250MG A1483868-50MG
76664-748EA 91.24 USD
76664-748 76664-750 76664-752
1-(Methoxymethyl)pyrimidine-2,4(1H,3H)-dione 97%
1-(Methoxymethyl)pyrimidine-2,4(1H,3H)-dione

Precaución: for Research Use Only

Formula: C₆H₈N₂O₃
MW: 156.1393 g/mol
CAS Number: 133625-42-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR