1-Pentylpyrimidine-2,4(1H,3H)-dione 97%

Supplier: Ambeed
Warning

A1483867-100MG A1483867-250MG
76664-744EA 53.05 USD
76664-744 76664-746
1-Pentylpyrimidine-2,4(1H,3H)-dione 97%
1-Pentylpyrimidine-2,4(1H,3H)-dione

Precaución: for Research Use Only

Formula: C₉H₁₄N₂O₂
MW: 182.2197 g/mol
CAS Number: 13350-87-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR