6-Butylpyrimidine-2,4(1H,3H)-dione 97%

Supplier: Ambeed
Warning

A1483864-100MG A1483864-1G A1483864-250MG
76664-738EA 20.16 USD
76664-738 76664-740 76664-742
6-Butylpyrimidine-2,4(1H,3H)-dione 97%
6-Butylpyrimidine-2,4(1H,3H)-dione

Precaución: for Research Use Only

Formula: C₈H₁₂N₂O₂
MW: 168.1931 g/mol
CAS Number: 116137-73-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR