ω-Heptalactam 96%

Supplier: Ambeed
Warning

Synonyms: 2-Azacyclooctanone, Azocan-2-one, ω-Heptanolactam, ω-Enantholactam

A1483803-100G A1483803-1G A1483803-25G A1483803-5G
76664-722EA 274.78 USD
76664-722 76664-724 76664-726 76664-728
ω-Heptalactam 96%
omega-Heptalactam

Precaución: for Research Use Only

Formula: C₇H₁₃NO
MW: 127.19 g/mol
MDL Number: MFCD00003271
CAS Number: 673-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR