(S)-2-(1-(tert-Butoxycarbonyl)pyrrolidin-3-yl)acetic acid ≥98%

Supplier: Ambeed
Warning

A1483791-1G A1483791-25G A1483791-5G
76664-716EA 25.47 USD
76664-716 76664-718 76664-720
(S)-2-(1-(tert-Butoxycarbonyl)pyrrolidin-3-yl)acetic acid ≥98%
(S)-2-(1-(tert-Butoxycarbonyl)pyrrolidin-3-yl)acetic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₄
MW: 229.2729 g/mol
MDL Number: MFCD05861546
CAS Number: 204688-61-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR