(S)-tert-Butyl 2-methyl-4-oxopiperidine-1-carboxylate ≥98%

Supplier: Ambeed
Warning

A1483785-1G A1483785-25G A1483785-5G
76664-684EA 27.59 USD
76664-684 76664-686 76664-688
(S)-tert-Butyl 2-methyl-4-oxopiperidine-1-carboxylate ≥98%
(S)-tert-Butyl 2-methyl-4-oxopiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₃
MW: 213.27 g/mol
MDL Number: MFCD09832895
CAS Number: 790667-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR