Methyl 2-((tert-butoxycarbonyl)amino)-3-iodopropanoate 98+%

Supplier: Ambeed
Warning

A1483783-1G A1483783-5G A1483783-25G
76664-676EA 55.17 USD
76664-676 77549-832 77624-388
Methyl 2-((tert-butoxycarbonyl)amino)-3-iodopropanoate 98+%
Methyl 2-((tert-butoxycarbonyl)amino)-3-iodopropanoate

Precaución: for Research Use Only

Formula: C₉H₁₆INO₄
MW: 329.13 g/mol
MDL Number: MFCD20231367
CAS Number: 889670-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR