(2R,6R)-2,6-Dimethylpiperazine dihydrochloride 98+%

Supplier: Ambeed
Warning

A1483779-100MG A1483779-1G A1483779-250MG A1483779-5G
76664-662EA 118.83 USD
76664-662 76664-664 76664-666 76664-668
(2R,6R)-2,6-Dimethylpiperazine dihydrochloride 98+%
(2R,6R)-2,6-Dimethylpiperazine dihydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₆Cl₂N₂
MW: 187.11 g/mol
MDL Number: MFCD13193063
CAS Number: 162240-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR