(R)-3-Methylpiperidine hydrochloride 98+%

Supplier: Ambeed
Warning

A1483778-100MG A1483778-250MG A1483778-1G A1483778-5G
76664-658EA 17 USD
76664-658 76664-660 77875-321 77875-322
(R)-3-Methylpiperidine hydrochloride 98+%
(R)-3-Methylpiperidine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₄ClN
MDL Number: MFCD22494946
CAS Number: 223792-48-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR