2-(2-Chloro-4-nitrophenyl)acetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-(2-Chloro-4-nitrophenyl)acetic Acid

A1483777-1G A1483777-25G
76664-656EA 88.06 USD
76664-656 77567-078
2-(2-Chloro-4-nitrophenyl)acetic acid 98%
2-(2-Chloro-4-nitrophenyl)acetic acid

Precaución: For research use only.

Formula: C₈H₆ClNO₄
MW: 215.591 g/mol
MDL Number: MFCD11036396
CAS Number: 73088-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR