4-Bromo-2-methyl-2H-indazole 98%

Supplier: Ambeed
Warning

A1483776-100G A1483776-1G A1483776-25G A1483776-5G
76664-648EA 1037.57 USD
76664-648 76664-650 76664-652 76664-654
4-Bromo-2-methyl-2H-indazole 98%
4-Bromo-2-methyl-2H-indazole

Precaución: for Research Use Only

Formula: C₈H₇BrN₂
MW: 211.0586 g/mol
MDL Number: MFCD09870046
CAS Number: 590417-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR