(R)-tert-Butyl 2-methyl-4-oxopiperidine-1-carboxylate 98+%
Supplier: Ambeed
|
Warning
|
A1483773-100G
A1483773-1G
A1483773-250MG
A1483773-25G
A1483773-5G
76664-638EA
2323.38
USD
76664-638
76664-640
76664-642
76664-644
76664-646
(R)-tert-Butyl 2-methyl-4-oxopiperidine-1-carboxylate 98+%
(R)-tert-Butyl 2-methyl-4-oxopiperidine-1-carboxylate
Precaución: for Research Use Only
Formula:
C₁₁H₁₉NO₃ MW: 213.27 g/mol |
MDL Number:
MFCD09832896 CAS Number: 790667-43-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...