2-Oxa-8-azaspiro[4.5]decane hydrochloride 98+%

Supplier: Ambeed
Warning

A1483770-250MG A1483770-25G A1483770-5G A1483770-100MG
76664-616EA 13 USD
76664-616 76664-618 76664-620 77875-317
2-Oxa-8-azaspiro[4.5]decane hydrochloride 98+%
2-Oxa-8-azaspiro[4.5]decane hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₅NO·ClH
MDL Number: MFCD11035930
CAS Number: 479195-19-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR