3-Fluoro-4-(hydroxymethyl)benzonitrile 98+%

Supplier: Ambeed
Warning

A1483759-100G A1483759-1G A1483759-25G A1483759-500G
76664-608EA 230 USD
76664-608 76664-610 76664-612 76664-614
3-Fluoro-4-(hydroxymethyl)benzonitrile 98+%
3-Fluoro-4-(hydroxymethyl)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₆FNO
MDL Number: MFCD09037779
CAS Number: 219873-06-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR