1-(2-Tetrahydropyranyl)-1H-pyrazole 98+%

Supplier: Ambeed
Warning

Synonyms: 1-(Tetrahydro-2H-pyran-2-yl)-1H-pyrazole

A1483753-100G A1483753-25G A1483753-500G A1483753-5G
76664-594EA 65.78 USD
76664-594 76664-596 76664-598 76664-600
1-(2-Tetrahydropyranyl)-1H-pyrazole 98+%
1-(2-Tetrahydropyranyl)-1H-pyrazole

Precaución: for Research Use Only

Formula: C₈H₁₂N₂O
MW: 152.20 g/mol
MDL Number: MFCD11044692
CAS Number: 449758-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR