tert-Butyl ((1S,2S)-2-hydroxycyclohexyl)carbamate 98%

Supplier: Ambeed
Warning

A1483749-1G A1483749-25G A1483749-5G
76664-580EA 33 USD
76664-580 76664-582 76664-584
tert-Butyl ((1S,2S)-2-hydroxycyclohexyl)carbamate 98%
tert-Butyl ((1S,2S)-2-hydroxycyclohexyl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₂₁NO₃
MDL Number: MFCD11042330
CAS Number: 145166-06-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR