Dibutyl vinylboronate 96% + (by GC) stabilised

Supplier: Ambeed
Warning

Synonyms: Vinylboronic Acid Dibutyl Ester, Dibutyl Vinylboronate

Stabilization: Stabilised with phenothiazine

A1483747-1G A1483747-250MG A1483747-25G A1483747-5G
76664-572EA 22 USD
76664-572 76664-574 76664-576 76664-578
Dibutyl vinylboronate 96% + (by GC) stabilised
Dibutyl vinylboronate

Precaución: for Research Use Only

Formula: C₁₀H₂₁BO₂
MW: 184.09 g/mol
MDL Number: MFCD00050049
CAS Number: 6336-45-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR