tert-Butyl ((1S,2S)-2-hydroxycyclopentyl)carbamate 99%

Supplier: Ambeed
Danger

A1483745-1G A1483745-25G A1483745-5G
76664-566EA 24 USD
76664-566 76664-568 76664-570
tert-Butyl ((1S,2S)-2-hydroxycyclopentyl)carbamate 99%
tert-Butyl ((1S,2S)-2-hydroxycyclopentyl)carbamate

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₃
MDL Number: MFCD11656038
CAS Number: 145106-43-0
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR