(4-Boc-1-piperazinium-1-ylmethyl)trifluoroborate internal salt 98+%

Supplier: Ambeed
Warning

A1483743-1G A1483743-25G A1483743-5G
76664-560EA 53 USD
76664-560 76664-562 76664-564
(4-Boc-1-piperazinium-1-ylmethyl)trifluoroborate internal salt 98+%
(4-Boc-1-piperazinium-1-ylmethyl)trifluoroborate internal salt

Precaución: for Research Use Only

Formula: C₁₀H₂₀BF₃N₂O₂
MDL Number: MFCD20134170
CAS Number: 1268340-97-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR