(4-(2-Hydroxypropan-2-yl)phenyl)boronic acid 98+%

Supplier: Ambeed
Warning

A1483741-100G A1483741-1G A1483741-25G A1483741-5G
76664-552EA 1686.84 USD
76664-552 76664-554 76664-556 76664-558
(4-(2-Hydroxypropan-2-yl)phenyl)boronic acid 98+%
(4-(2-Hydroxypropan-2-yl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₃BO₃
MW: 180.009 g/mol
MDL Number: MFCD09992905
CAS Number: 886593-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR