Potassium (tert-butoxymethyl)trifluoroborate 98+%

Supplier: Ambeed
Warning

Synonyms: potassium [(tert-butoxy)methyl]trifluoroboranuide, Potassium (tert-butoxymethyl)trifluoroborate, Potassium (tert-butoxymethyl)(trifluoro)borate(1-)

A1483729-1G A1483729-25G A1483729-5G
76664-530EA 30 USD
76664-530 76664-532 76664-534
Potassium (tert-butoxymethyl)trifluoroborate 98+%
Potassium (tert-butoxymethyl)trifluoroborate

Precaución: for Research Use Only

Formula: C₅H₁₁BF₃KO
MW: 194.046 g/mol
MDL Number: MFCD11505924
CAS Number: 910251-10-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR