3-Hydroxyphenylboronic acid pinacol ester 98+%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
A1483725-1G
A1483725-5G
A1483725-100G
76664-526EA
7
USD
76664-526
76664-528
77624-380
3-Hydroxyphenylboronic acid pinacol ester 98+%
3-Hydroxyphenylboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₂H₁₇BO₃ |
MDL Number:
MFCD02093755 CAS Number: 214360-76-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...