2-Methyl-5,7-dihydrothieno[3,4-d]pyrimidine 98+%

Supplier: Ambeed
Warning

A1483655-100MG A1483655-1G A1483655-250MG
76664-510EA 6 USD
76664-510 76664-512 76664-514
2-Methyl-5,7-dihydrothieno[3,4-d]pyrimidine 98+%
2-Methyl-5,7-dihydrothieno[3,4-d]pyrimidine

Precaución: for Research Use Only

Formula: C₇H₈N₂S
MDL Number: MFCD00038619
CAS Number: 36267-71-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR