1-Bromotetradecane 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
Myristyl bromide
A1483013-100G
A1483013-500G
A1483013-1000G
76664-494EA
16
USD
76664-494
76664-496
77597-558
1-Bromotetradecane 99%
1-Bromotetradecane
Precaución: for Research Use Only
Formula:
BrH₂C(CH₂)₁₂CH₃ |
MDL Number:
MFCD00000228 CAS Number: 112-71-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...