Thiodiglycolic anhydride 99%

Supplier: Ambeed
Warning

Synonyms: 1,4-Oxathiane-2,6-dione

A1483003-1000G A1483003-100G A1483003-25G A1483003-500G
76664-486EA 262.05 USD
76664-486 76664-488 76664-490 76664-492
Thiodiglycolic anhydride 99%
Thiodiglycolic anhydride

Precaución: for Research Use Only

Formula: C₄H₄O₃S
MW: 132.13 g/mol
MDL Number: MFCD00051689
CAS Number: 3261-87-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR