1-Chloro-2-phenylethane 99%

Supplier: Ambeed
Warning

Synonyms: 2-Phenylethyl chloride, (2-Chloroethyl)benzene, 2-(Chloroethyl)benzene

A1482993-100G A1482993-25G A1482993-500G A1482993-1000G
76664-466EA 20.16 USD
76664-466 76664-468 76664-470 77624-362
1-Chloro-2-phenylethane 99%
1-Chloro-2-phenylethane

Precaución: for Research Use Only

Formula: ClCH₂CH₂C₆H₅
MW: 140.61 g/mol
MDL Number: MFCD00000977
CAS Number: 622-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR