1-Decanethiol 99%

Supplier: Ambeed
Danger

Synonyms: n-Decanethiol, 1-Mercaptodecane, n-Decyl mercaptan

A1482987-25G A1482987-100G
76664-464EA 18 USD
76664-464 77592-790
1-Decanethiol 99%
1-Decanethiol

Precaución: for Research Use Only

Formula: C₁₀H₂₂S
MW: 174.35 g/mol
MDL Number: MFCD00004884
CAS Number: 143-10-2
UN: 3334
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR