2-Ethylhexyl acetate 99+%

Supplier: Ambeed
Warning

A1482983-25G A1482983-500G
76664-462EA 6 USD
76664-462 77875-297
2-Ethylhexyl acetate 99+%
2-Ethylhexyl acetate

Precaución: for Research Use Only

Formula: C₁₀H₂₀O₂
MDL Number: MFCD00027249
CAS Number: 103-09-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR