Methyl-5-amino-2-fluorobenzenecarboxylate 98+%

Supplier: Ambeed
Warning

A1482921-1G A1482921-25G A1482921-5G
76664-440EA 22 USD
76664-440 76664-442 76664-444
Methyl-5-amino-2-fluorobenzenecarboxylate 98+%
Methyl-5-amino-2-fluorobenzenecarboxylate

Precaución: for Research Use Only

Formula: C₈H₈FNO₂
MDL Number: MFCD09037918
CAS Number: 56741-34-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR