2,5-Dioxopyrrolidin-1-yl nicotinate 97%

Supplier: Ambeed
Warning

A1482897-1G A1482897-250MG A1482897-25G A1482897-5G
76664-432EA 39.26 USD
76664-432 76664-434 76664-436 76664-438
2,5-Dioxopyrrolidin-1-yl nicotinate 97%
2,5-Dioxopyrrolidin-1-yl nicotinate

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O₄
MW: 220.1815 g/mol
MDL Number: MFCD00619111
CAS Number: 78348-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR