4,4'-((6-((2-Aminoethyl)amino)-1,3,5-triazine-2,4-diyl)bis(azanediyl))bis(butan-1-ol) 97%

Supplier: Ambeed
Warning

A1482821-100MG A1482821-50MG
76664-428EA 239.77 USD
76664-428 76664-430
4,4'-((6-((2-Aminoethyl)amino)-1,3,5-triazine-2,4-diyl)bis(azanediyl))bis(butan-1-ol) 97%
4,4'-((6-((2-Aminoethyl)amino)-1,3,5-triazine-2,4-diyl)bis(azanediyl))bis(butan-1-ol)

Precaución: for Research Use Only

Formula: C₁₃H₂₇N₇O₂
MW: 313.3992 g/mol
CAS Number: 2757730-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR