N-(3,5-Dichloro-4-iodophenyl)acetamide 97+%

Supplier: Ambeed
Warning

A1482803-100MG A1482803-1G A1482803-250MG A1482803-5G
76664-420EA 25.47 USD
76664-420 76664-422 76664-424 76664-426
N-(3,5-Dichloro-4-iodophenyl)acetamide 97+%
N-(3,5-Dichloro-4-iodophenyl)acetamide

Precaución: for Research Use Only

Formula: C₈H₆Cl₂INO
MW: 329.9498 g/mol
CAS Number: 89894-32-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR