N-Benzyl-N,N-dimethyl-2-(2-(2-methyl-4-(2,4,4-trimethylpentan-2-yl)phenoxy)ethoxy)ethanaminium chloride 92+% (by HPLC)

Supplier: Ambeed
Warning

A1482801-100MG A1482801-1G A1482801-250MG
76664-414EA 12 USD
76664-414 76664-416 76664-418
N-Benzyl-N,N-dimethyl-2-(2-(2-methyl-4-(2,4,4-trimethylpentan-2-yl)phenoxy)ethoxy)ethanaminium chloride 92+% (by HPLC)
N-Benzyl-N,N-dimethyl-2-(2-(2-methyl-4-(2,4,4-trimethylpentan-2-yl)phenoxy)ethoxy)ethanaminium chloride

Precaución: for Research Use Only

Formula: C₂₈H₄₄ClNO₂
MDL Number: MFCD00011821
CAS Number: 101912-16-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR