1,16-Hexadecanedioic acid 98%

Supplier: Ambeed
Warning

Synonyms: 1,14-Tetradecanedicarboxylic acid, Thapsic acid, Hexadecanedioic acid

A1482669-500G A1482669-5G A1482669-25G A1482669-100G
76664-394EA 214 USD
76664-394 76664-396 77597-554 77875-294
1,16-Hexadecanedioic acid 98%
1,16-Hexadecanedioic acid

Precaución: for Research Use Only

Formula: C₁₆H₃₀O₄
MDL Number: MFCD00002746
CAS Number: 505-54-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR