1,11-Dibromoundecane ≥98%

Supplier: Ambeed
Warning

Synonyms: Undecamethylene dibromide

A1482661-100G A1482661-25G A1482661-5G
76664-382EA 344.8 USD
76664-382 76664-384 76664-386
1,11-Dibromoundecane ≥98%
1,11-Dibromoundecane

Precaución: for Research Use Only

Formula: BrH₂C(CH₂)₉CH₂Br
MW: 314.1 g/mol
MDL Number: MFCD00000224
CAS Number: 16696-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR