Undecane-1,11-diol 98+%

Supplier: Ambeed
Warning

A1482659-100G A1482659-500G A1482659-25G
76664-378EA 19 USD
76664-378 76664-380 77597-552
Undecane-1,11-diol 98+%
Undecane-1,11-diol

Precaución: for Research Use Only

Formula: C₁₁H₂₄O₂
MDL Number: MFCD00041568
CAS Number: 765-04-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR