4-Methoxypyridin-2(1H)-one 98%

Supplier: Ambeed
Warning

A1482546-1G A1482546-250MG A1482546-5G
76664-366EA 83 USD
76664-366 76664-368 76664-370
4-Methoxypyridin-2(1H)-one 98%
4-Methoxypyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₆H₇NO₂
MDL Number: MFCD09839191
CAS Number: 52545-13-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR