ω,3-Dibromoacetophenone 98+%

Supplier: Ambeed
Danger

A1482514-1G A1482514-25G A1482514-5G A1482514-100G
76664-354EA 8 USD
76664-354 76664-356 76664-358 77622-248
ω,3-Dibromoacetophenone 98+%
ω,3-Dibromoacetophenone

Precaución: for Research Use Only

Formula: C₈H₆Br₂O
MDL Number: MFCD00082698
CAS Number: 18523-22-3
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR