4-(Trifluoromethoxy)phenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 4,4,5,5-Tetramethyl-2-(4-(trifluoromethoxy)phenyl)-1,3,2-dioxaborolane

A1482496-1G A1482496-25G A1482496-5G
76664-340EA 7.43 USD
76664-340 76664-342 76664-344
4-(Trifluoromethoxy)phenylboronic acid pinacol ester 98%
4-(Trifluoromethoxy)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₆BF₃O₃
MW: 288.0706 g/mol
MDL Number: MFCD05863920
CAS Number: 474709-28-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR