Boc-Tyr(Boc)-OH 98+%

Supplier: Ambeed
Warning

A1482483-100G A1482483-1G A1482483-25G A1482483-5G
76664-332EA 459 USD
76664-332 76664-334 76664-336 76664-338
Boc-Tyr(Boc)-OH 98+%
Boc-Tyr(Boc)-OH

Precaución: for Research Use Only

Formula: C₁₉H₂₇NO₇
MDL Number: MFCD00038856
CAS Number: 20866-48-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR