(S)-Ethyl 2-acetamido-4-methylpentanoate 98+%

Supplier: Ambeed
Warning

A1482477-100G A1482477-1G A1482477-25G A1482477-5G
76664-316EA 395 USD
76664-316 76664-318 76664-320 76664-322
(S)-Ethyl 2-acetamido-4-methylpentanoate 98+%
(S)-Ethyl 2-acetamido-4-methylpentanoate

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₃
MDL Number: MFCD09952004
CAS Number: 4071-36-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR