(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)pentanedioic acid 98+%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Glu-OH, N-Fmoc-L-glutamic Acid

A1482468-100G A1482468-25G A1482468-500G A1482468-5G
76664-310EA 44 USD
76664-310 76664-312 76664-314 77875-278
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)pentanedioic acid 98+%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)pentanedioic acid

Precaución: for Research Use Only

Formula: C₂₀H₁₉NO₆
MDL Number: MFCD00237657
CAS Number: 121343-82-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR