H-Asp(OMe)-OH 98+%

Supplier: Ambeed
Warning

Synonyms: (2S)-2-Amino-4-methoxy-4-oxo-butanoic acid, 4-Methyl hydrogen L-aspartate

A1482465-25G A1482465-5G
76664-306EA 32.89 USD
76664-306 76664-308
H-Asp(OMe)-OH 98+%
H-Asp(OMe)-OH

Precaución: for Research Use Only

Formula: C₅H₉NO₄
MW: 147.13 g/mol
MDL Number: MFCD00055775
CAS Number: 2177-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR