(S)-3-([1,1'-Biphenyl]-4-yl)-2-aminopropanoic acid 98%

Supplier: Ambeed
Warning

Synonyms: L-Biphenylalanine, H-4-Phenyl-Phe-OH

A1482462-100MG A1482462-1G A1482462-250MG A1482462-5G
76664-292EA 7.43 USD
76664-292 76664-294 76664-296 76664-298
(S)-3-([1,1'-Biphenyl]-4-yl)-2-aminopropanoic acid 98%
(S)-3-([1,1'-Biphenyl]-4-yl)-2-aminopropanoic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₅NO₂
MW: 241.29 g/mol
MDL Number: MFCD01318878
CAS Number: 155760-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR