S-(Trifluoromethyl) [1,1'-biphenyl]-4-carbothioate 98%

Supplier: Ambeed
Warning

A1482322-1G A1482322-250MG A1482322-5G A1482322-25G
76664-264EA 79 USD
76664-264 76664-266 76664-268 77875-274
S-(Trifluoromethyl) [1,1'-biphenyl]-4-carbothioate 98%
S-(Trifluoromethyl) [1,1'-biphenyl]-4-carbothioate

Precaución: for Research Use Only

Formula: C₁₄H₉F₃OS
CAS Number: 1927969-10-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR