2-(4-Isopropylphenoxy)acetic acid 95% (by HPLC)
Supplier: Ambeed
|
Warning
|
A1482263-100G
A1482263-5G
76664-250EA
144
USD
76664-250
76664-252
2-(4-Isopropylphenoxy)acetic acid 95% (by HPLC)
2-(4-Isopropylphenoxy)acetic acid
Precaución: for Research Use Only
Formula:
C₁₁H₁₄O₃ |
MDL Number:
MFCD00014364 CAS Number: 1643-16-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...