tert-Butyl 3-(2-(2-(2-iodoethoxy)ethoxy)ethoxy)propanoate 95%

Supplier: Ambeed
Warning

A1481919-1G A1481919-250MG A1481919-100MG
76664-242EA 205.82 USD
76664-242 77597-524 77624-356
tert-Butyl 3-(2-(2-(2-iodoethoxy)ethoxy)ethoxy)propanoate 95%
tert-Butyl 3-(2-(2-(2-iodoethoxy)ethoxy)ethoxy)propanoate

Precaución: for Research Use Only

Formula: C₁₃H₂₅IO₅
MW: 388.2391 g/mol
MDL Number: MFCD31656966
CAS Number: 1291090-50-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR