tert-Butyl 3-(2-(2-iodoethoxy)ethoxy)propanoate 97%

Supplier: Ambeed
Warning

Synonyms: tert-butyl 3-[2-(2-iodoethoxy)ethoxy]propanoate

A1481830-1G A1481830-5G A1481830-250MG
76664-238EA 173.99 USD
76664-238 76664-240 77549-826
tert-Butyl 3-(2-(2-iodoethoxy)ethoxy)propanoate 97%
tert-Butyl 3-(2-(2-iodoethoxy)ethoxy)propanoate

Precaución: for Research Use Only

Formula: C₁₁H₂₁IO₄
MW: 344.1865 g/mol
MDL Number: MFCD31656965
CAS Number: 1938057-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR