2,5-Dioxopyrrolidin-1-yl 3-(2-(prop-2-yn-1-yloxy)ethoxy)propanoate 96+%
Supplier: Ambeed
|
Warning
|
Synonyms:
Propargyl-PEG2-NHS ester
A1481819-1G
A1481819-250MG
A1481819-100MG
A1481819-5G
76664-236EA
190
USD
76664-236
77597-522
77624-354
77875-261
2,5-Dioxopyrrolidin-1-yl 3-(2-(prop-2-yn-1-yloxy)ethoxy)propanoate 96+%
2,5-Dioxopyrrolidin-1-yl 3-(2-(prop-2-yn-1-yloxy)ethoxy)propanoate
Precaución: for Research Use Only
Formula:
C₁₂H₁₅NO₆ |
CAS Number:
2512228-06-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...