Fmoc-β-(3-benzothienyl)-Ala-OH 98+%
Supplier: Ambeed
|
Warning
|
Synonyms:
Fmoc-3-Ala(3-benzothienyl)-OH, Fmoc-β-(3-benzothienyl)-Ala-OH
A1481680-100MG
A1481680-1G
A1481680-250MG
A1481680-25G
A1481680-5G
76664-214EA
11
USD
76664-214
76664-216
76664-218
76664-220
76664-222
Fmoc-β-(3-benzothienyl)-Ala-OH 98+%
Fmoc-beta-(3-benzothienyl)-Ala-OH
Precaución: for Research Use Only
Formula:
C₂₆H₂₁NO₄S |
MDL Number:
MFCD00672562 CAS Number: 177966-60-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...